Learn More
Thermo Scientific™ Remel™ Calgiswab™ Type 2 Swab Collection and Transport System, Wood-shaft
Description
Thermo Scientific™ Remel Calgiswab Swab Collection and Transport System is designed for nasopharyngeal specimen collection.
Specifications
Specifications
| Product Type | Remel Calgiswab |
| Format | Swab |
| Description | Wood-shaft; 1/Envelope; 50/Pk. |
| For Use With (Application) | Healthcare |
| Packaging | 1/Envelope |
| Quantity | 50/Pk. |
| Shaft Material | Wood |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.