Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Cary-Blair Transport Medium
GSA_VA

Catalog No. R060450
Change view
Click to view available options
Quantity:
20 x 9 mL
100 x 9 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060450 100 x 9 mL
R060452 20 x 9 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R060450 Supplier Thermo Scientific™ Supplier No. R060450
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel Cary-Blair Transport Medium is for the transport of clinical specimens, primarily stool and rectal swabs.

Thermo Scientific™ Remel Cary-Blair Transport Medium is for the transport of clinical specimens, primarily stool and rectal swabs.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Cary-Blair Transport Medium
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 100 x 9 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.