Learn More
Thermo Scientific™ Remel™ Cetrimide Selective Agar (Dehydrated)
Catalog No. R452802 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R452802 500 g
1 options
Catalog No. R452802 Supplier Thermo Scientific™ Supplier No. R452802
Add to Cart
Add to Cart
Selectively isolate and presumptively identify Pseudomonas aeruginosa and other gram-negative, non-fermentative bacteria using this medium.
Selectively isolate and presumptively identify Pseudomonas aeruginosa and other non-fermentative, gram-negative bacilli using Thermo Scientific™ Remel™ Cetrimide Selective Agar (Dehydrated). King et al. developed a medium called Tech Agar which was used to enhance pyocyanin production by Pseudomonas1. In 1955, Lowbury described the use of cetrimide in a selective medium for P. aeruginosa2. This formulation is recommended by the United States Pharmacopeia (USP) and the AOAC International (AOAC)3,4. Centrimide Agar has the formula of Tech Agar but is modified by the addition of centrimide.
Selectively isolate and presumptively identify Pseudomonas aeruginosa and other non-fermentative, gram-negative bacilli using Cetrimide Selective Agar (Dehydrated).
  • Selective: Centrimide in the medium inhibits bacteria other than P. aeruginosa by causing nitrogen and to be released from bacterial cells, making the medium selective for P. aeruginosa.
  • Easy-to-read: P. aeruginosa forms fluorescent green colonies which can be easily differentiated from the other non-fluoroscent colonies.
  • This formulation is recommended by the United States Pharmacopeia (USP) and the AOAC International (AOAC)3,4. Centrimide Agar has the formula of Tech Agar but is modified by the addition of centrimide.
This medium contains potassium sulfate and magnesium chloride which stimulate the production of pyocyanin and fluorescein. Fluorescein production is visualized using an ultraviolet light. P. aeruginosa is characterized by production of pyocyanin, a blue-green, water-soluble, non-fluorescent phenazine pigment. Centrimide (N-acetyl-NNN-trimethyl-ammonium bromide, cetavlon) causes nitrogen and phosphorus to be released from bacterial cells and inhibits bacteria other than P. aeruginosa.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM, USP.
Description Cetrimide Selective Agar
Form Powder
Product Type Selective Medium
Quantity 500 g
REF R110131 and REF R112040 not intended for IVD use.