Learn More
Thermo Scientific™ Remel™ Color Slide™ RF Rapid Latex Test
Catalog No. 23033138A
Change view
Click to view available options
100 tests
40 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-033-138A 40 tests
23-033-138B 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-033-138A Supplier Thermo Scientific™ Supplier No. R2471328
Includes
Positive and negative controls, latex reagent, sample diluent, pipets, and disposable test cards
Latex agglutination slide test for the qualitative and semi-quantitative determination of circulating rheumatoid factor (RF) in human serum
  • CLIA Complexity: Non-Waived
  • CPT Codes: 86431
  • Utilizes mono-dispersed latex reagent
  • Semi-quantitative titers can be performed directly on the reaction card for efficiency and accuracy
  • No sample dilution is required for qualitative procedure
  • Positive control is assayed to assist in regulatory compliance to prove proficiency of titering techniques
  • Results in uniformly smooth negatives
  • Test results within one minute
  • For in vitro diagnostic use