Learn More
Thermo Scientific™ Remel™ Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80
Description
Thermo Scientific™ Remel Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80 is used for the cultivation of bacteria, yeast and fungi.
- Manufactured in a clean room environment to provide the highest level of sterility assurance with no compromise of growth characteristics
- Each lot is tested to ensure consistent, reproducible results
- Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifications
Specifications
| Description | Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80 |
| Format | Contact - Single Bag |
| Product Type | Agar Plate |
| Quantity | 100/Pk. |
Not intended for IVD use.