Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cornmeal Agar with Polysorbate 80 with or without Trypan Blue
GSA_VA

Catalog No. R09290
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
20mm x 150mm Pour Tube
Monoplate Deep Pour
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R09290 20mm x 150mm Pour Tube
R01328 12mm x 85mm Monoplate
R01354 Monoplate Deep Pour
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R09290 Supplier Thermo Scientific™ Supplier No. R09290
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cultivate fungi and stimulate chlamydospore production by Candida albicans with this ready-to-use medium.

Cultivate fungi and stimulate chlamydospore production by Candida albicans with Thermo Scientific™ Remel Cornmeal Agar with Polysorbate 80 general purpose medium, with or without Trypan Blue. Polack and Benham demonstrated the use of cornmeal agar for studying differential mycelial characteristics of Candida species1,2. Adding polysorbate 80 to cornmeal agar stimulates chlamydospore formation by Candida albicans3-4.

Cornmeal Agar with Polysorbate 80 with or without trypan blue is a solid medium recommended for use in qualitative procedures as a general-purpose media for cultivating fungi and stimulating chlamydospore production by Candida albicans.

  • General-purpose media: cultivate fungi and stimulate chlamydospore formation by Candida albicans.
  • Easy-to-read: Trypan blue provides a contrasting background for the optimal visualization of chlamydospores.
  • Ready-to-use: the convenience of prepared media.

Cornmeal agar consists of an infusion of cornmeal and agar, which provides the essential nutrients needed to support the growth of fungi and the production of chlamydospores by Candida albicans. The agar is a solidifying agent. Polysorbate 80 reduces surface tension and enhances the development of chlamydospores. Trypan blue is an optimal additive that provides a contrasting background, thereby permitting optimal visualization of chlamydospores.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Cornmeal Agar w/Tween™ 80
Format 20mm x 150mm Pour Tube
Product Type Agar Plate
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.