Learn More
Thermo Scientific™ D/E (Dey-Engley) Neutralizing Broth (Dehydrated)
Catalog No. R453042 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453042 500 g
1 options
Catalog No. R453042 Supplier Thermo Scientific™ Supplier No. R453042
Add to Cart
Edge
Add to Cart
Neutralize chemical disinfectants to confirm bacteriostatic and bactericidal activity in environmental sampling procedures with D/E Neutralizing Broth.
Neutralize chemical disinfectants to distinguish bacteriostatic and bactericidal activity in environmental sampling procedures using Thermo Scientific™ Remel™ D/E (Dey-Engley) Neutralizing Broth (Dehydrated). This medium was formulated by Engley and Dey in 1970 for neutralizing and testing antiseptics and disinfectants1. It is recommended in the Compendium of Methods for the Microbiological Examination of Foods by the American Public Health Association (APHA) for environmental sampling2.
Neutralize chemical disinfectants to distinguish bacteriostatic and bactericidal activity in environmental sampling procedures using D/E (Dey-Engley) Neutralizing Broth. The medium can neutralize a range of chemicals including mercurial, iodine or chlorine preparations, quaternary ammonium compounds, formaldehyde and glutaraldehyde.
  • Easy to Identify: Bromocresol Purple is used as an indicator for the fermentation of dextrose by shifting from purple to yellow at acidic pH.
  • Easy to differentiate: Subculture all tubes of D/E Neutralizing Broth to appropriate media such as D/E Agar to observe growth (viable microorganisms).
    • Bacteriostatic substance – Growth from negative D/E Neutralizing Broth tubes.
    • Bactericidal substance – No growth from negative D/E Neutralizing Broth tubes.
  • Recommended in the Compendium of Methods for the Microbiological Examination of Foods by the American Public Health Association (APHA) for environmental sampling2.
This medium contains peptone and yeast extract that provide a source of nitrogen, carbon, and vitamins. Dextrose provides energy and sodium thiosulfate is incorporated to neutralize chlorine and iodine. Polysorbate 80 neutralizes phenols and formalin. Sodium thioglycolate neutralizes mercurials. Sodium bisulfite neutralizes formaldehyde and glutaraldehyde. Lecithin neutralizes quaternary ammonium compounds.. This medium is suitable for environmental sampling where neutralization of the chemical is important to determine its bactericidal activity.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description D/E Neutralizing Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Not intended for IVD use.