Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ D/E (Dey-Engley) Neutralizing Broth

Catalog No. R060770
Change view
Click to view available options
Format:
15mm x 103mm Tube
20mm x 150mm Tube
WMB Lug
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R060770 15mm x 103mm Tube
R09334 20mm x 150mm Tube
R112160 WMB Lug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R060770 Supplier Thermo Scientific™ Supplier No. R060770
Add to Cart
Add to Cart
This item is not returnable. View return policy

Neutralize chemical disinfectants to confirm bacteriostatic and bactericidal activity in environmental sampling procedures with D/E Neutralizing Broth.

Neutralize chemical disinfectants to distinguish bacteriostatic and bactericidal activity in environmental sampling procedures using Thermo Scientific™ Remel™ D/E (Dey-Engley) Neutralizing Broth. This medium was formulated by Engley and Dey in 1970 for neutralizing and testing antiseptics and disinfectants1. It is recommended in the Compendium of Methods for the Microbiological Examination of Foods by the American Public Health Association (APHA) for environmental sampling2.

Neutralize chemical disinfectants to distinguish bacteriostatic and bactericidal activity in environmental sampling procedures using D/E (Dey-Engley) Neutralizing Broth. The medium can neutralize a range of chemicals including mercurial, iodine or chlorine preparations, quaternary ammonium compounds, formaldehyde and glutaraldehyde.

  • Ready to Use: Convenience of prepared media.
  • Easy to Identify: As dextrose is utilized by the bacteria the pH indicator changes from a purple to yellow color.
  • Easy to differentiate: Subculture all tubes of D/E Neutralizing Broth to appropriate media such as D/E Agar to observe growth (viable microorganisms).
    • Bacteriostatic substance – Growth from negative D/E Neutralizing Broth tubes.
    • Bactericidal substance – No growth from negative D/E Neutralizing Broth tubes.
  • Recommended in the Compendium of Methods for the Microbiological Examination of Foods by the American Public Health Association (APHA) for environmental sampling.

This medium contains peptone and yeast extract that provide nutrients such as amino acids, peptides, nitrogenous compounds, and B-complex vitamins which are necessary for growth. Dextrose provides energy, lecithin neutralizes quaternary ammonium compounds, polysorbate 80 neutralizes phenolics, thioglycolate neutralizes mercurials, sodium thiosulfate neutralizes iodine and chlorine preparations and sodium bisulfate neutralizes formaldehyde and glutaraldehyde. This medium is suitable for environmental sampling where neutralization of the chemical is important to determine its bactericidal activity.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: COMPF.
Description D/E Neutralizing Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 5 mL

Not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.