Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Demi-Fraser Broth Base

Catalog No. R453462 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453462 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R453462 Supplier Thermo Scientific™ Supplier No. R453462
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Demi-Fraser Broth Base (Dehydrated) is recommended for use in qualitative procedures for the selective isolation and cultivation of Listeria monocytogenes.

Selectively isolate and cultivate Listeria monocytogenes from food and environmental samples using Thermo Scientific™ Remel™ Demi-Fraser Broth Base (Dehydrated).

Demi-Fraser Broth Base is a modification of Fraser Broth Base developed by Fraser and Sperber and used for rapid detection of Listeria in food and environmental samples1. The nalidixic acid and acriflavine concentrations in this medium have been reduced to 10 mg/L and 12.5 mg/L respectively in accordance with L'association française de normalisation (AFNOR) guidelines2.

Listeria monocytogenes is a common contaminant in raw milk, meats, vegetables, seafood and in the food processing environment3,4. Isolation of Listeria species presents a grave challenge because it can grow in pH environments ranging from 5.0 – 9.6 and even under refrigeration.

Successful recovery of Listeria from food and environmental sources requires a medium which can promote the growth of potentially injured cells while inhibiting the growth of non-Listeria background organisms. The isolation of low numbers of L. monocytogenes can be improved by enrichment procedures using a broth medium.

  • Selective Medium: Contains lithium chloride, nalidixic acid and acriflavine
    Lithium chloride – inhibits most enterococci which can also hydrolyse esculin.
    Nalidixic acid – blocks DNA replication of susceptible bacteria.
    Acriflavine – inhibits growth of secondary gram-positive microflora.
  • Easy-to-interpret: Color change on visual inspection - blackening of the solution
  • Conforms to L’association française de normalisation (AFNOR) guidelines

Demi-Fraser Broth Base contains casein peptone, meat peptone and beef extract that provide essential amino acids, peptides, and nitrogen necessary for growth of Listeria species. Yeast extract supplies B-complex vitamins, sodium chloride provides essential electrolytes and maintains osmotic equilibrium. Disodium phosphate and monosodium phosphate act as buffers and maintain the pH. Lithium chloride, nalidixic acid, and acriflavine act as selective agents. 0.1 mL of Fraser Broth Additive (REF R450361) is added to every 10 mL of prepared Demi-Fraser Broth Base immediately before use. This supplies the ferric ions in form of ferric ammonium citrate. All Listeria species hydrolyse esculin to form 6,7-dihydroxycoumarin, which combines with ferric ions to blacken the medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Demi-Fraser Broth Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.