Description
Selectively isolate and enrich Enterobacteriaceae from food and animal feeds with EE Broth Mossel.
Enrich and selectively isolate Enterobacteriaceae with Thermo Scientific™ Remel™ EE Broth Mossel (Dehydrated).
Traditional methods to monitor quality of foods and animal feeds tested for the presence of coliforms as indicator organisms. Mossel et al. expanded on these traditional methods by examining food products for all Enterobacteriaceae including glucose-positive, lactose-negative gram-negative bacilli (e.g. Salmonella)2-4. Early formulations contained bile salt variations that inhibited some indicator organisms. EE Broth Mossel contains purified ox bile (oxgall) instead of bile salts.
EE Broth Mossel is used for the selective enrichment of Enterobacteriaceae in the bacteriological examination of foods. It contains purified ox bile (oxgall) in place of bile salts and disodium phosphate to improve the buffering capacity of the medium and encourage early growth of indicator organisms.
- Selective: Brilliant green dye and oxgall inhibit gram-positive organisms
- Recommended by the American Public Health Association for most probable number assay and as an enrichment broth in straight enrichment procedures5.
- Formulated in conformance with harmonized United States and European Pharmocopoeia guidelines for the detection of bile-tolerant, gram-negative bacteria6,7.
Peptone provides nitrogen, carbon and sulfur compounds essential for bacterial growth. Dextrose serves as the energy source. Disodium phosphate and monopotassium phosphate act as buffering agents. Brilliant green and oxgall are selective agents which inhibit gram-positive organisms.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | EE Broth Mossel |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |