Learn More
Thermo Scientific™ Remel™ Fluid A
Description
Thermo Scientific™ Remel Fluid A with 0.1% peptone (Peptone Fluid) is used as diluting or rinsing fluid for sterility testing.
Specifications
Specifications
| Description | Fluid A |
| Format | BB Septum - 300mL |
| Product Type | Liquid Medium |
| Quantity | 10 x 300 mL |
Not intended for IVD use.