Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fluid Thioglycollate Medium (Dehydrated)

Catalog No. R453452 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453452 500 g
R453456 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R453452 Supplier Thermo Scientific™ Supplier No. R453452
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate aerobes and anaerobes and carry out sterility testing using Fluid Thioglycollate Medium.

Cultivate aerobes and anaerobes and carry out sterility testing using Thermo Scientific™ Remel™ Fluid Thioglycollate Medium (Dehydrated). This general-purpose medium was first described by Brewer in 19401 consisting of a small amount of agar and a reducing substance to initiate the growth of anaerobes2 and for sterility testing. This medium is recommended by the United States Pharmacopeia (USP) and the AOAC International (AOAC), the American Public Health Association (APHA) and Food and Drug Administration (FDA) in the Bacteriological Analytical Manual Online3-6.

Cultivate aerobes and anaerobes using Fluid Thioglycollate Medium.

  • Recommended by APHA, AOAC, USP and FDA.
  • Ease of Identification: Resazurin is an oxidation-reduction indicator which turns pink when oxidation is increased.

This medium is used for qualitative procedures. This medium contains casein peptone and cystine that provides nitrogen and carbon compounds, agar that impedes oxygen diffusion, and sodium thioglycollate, which is a reducing agent that maintains a low oxygen tension in anaerobic medium. Yeast extract supplies B-complex vitamins and enhances the growth of microorganisms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: USP
Description Fluid Thioglycollate Medium
Form Powder
Media Type Fluid Thioglycollate Medium
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.