Learn More
Thermo Scientific™ Remel™ Fraser Broth Base
Catalog No. R453482
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453482 500 g
1 options
This item is not returnable. View return policy
Catalog No. R453482 Supplier Thermo Scientific™ Supplier No. R453482
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use with acriflavin, nalidixic acid, and ferric ammonium citrate for the selective enrichment of Listeria species.
Remel Fraser Broth Base is for use with acriflavin, nalidixic acid, and ferric ammonium citrate for the selective enrichment of Listeria species.
Also available: Fraser Broth Additive, Part No. 450361.
TRUSTED_SUSTAINABILITY
Specifications
Description Fraser Broth Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
For Laboratory Use Only