Learn More
Thermo Scientific™ Remel™ GC Agar Base
Catalog No. R453506 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Form:
Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
R453506 10 kg -
R453502 500 g Powder
R453504 2.5 kg Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R453506 Supplier Thermo Scientific™ Supplier No. R453506
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use with hemoglobin and GCHI enrichment for the isolation and cultivation of Neisseria/gonorrhoeae and other fastidious microorganisms.
Remel GC Agar Base is used with hemoglobin and GCHI enrichment for the isolation and cultivation of Neisseria/gonorrhoeae and other fastidious microorganisms.
Also available: GCHI Enrichment with Rehydrating Solution, Part No. R450411; VCN Selective Supplement, Part No. R450901; and VCNT Selective Supplement, Part No. R450921.
TRUSTED_SUSTAINABILITY
Specifications
Description GC Agar Base
Format Powder
Media Type GC Agar Base
Product Type Nutrient Agar
Quantity 10 kg
For Laboratory Use Only