Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GN Broth (Dehydrated)

Catalog No. R453512 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453512 500 g
R453514 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R453512 Supplier Thermo Scientific™ Supplier No. R453512
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selectively enrich gram-negative bacilli, especially Salmonella and Shigella spp. using GN Broth.

Selectively enrich gram-negative bacilli, especially Salmonella and Shigella spp. using Thermo Scientific™ Remel™ GN Broth (Dehydrated). It was developed by Hanja as an enrichment medium which has proven superior to direct inoculation of media for recovery and enrichment of enteric gram-negative bacilli1. Taylor and Schelhart found GN Broth to be more effective than selenite broth for recovery of Shigella spp2.

Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.

Selectively enrichgram-negative bacilli, especially Salmonella and Shigella spp. using GN Broth.

This medium is a ready-to-use liquid medium used for qualitative procedures. This medium contains casein and meat peptone that provide amino acids and other nitrogenous compounds to support bacterial growth. Dextrose and mannitol are energy sources and their concentrations are balanced to limit the growth of Proteus and encourage the growth of Salmonella and Shigella. Sodium deoxycholate and sodium citrate inhibit gram-positive organisms.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: USDA.
Description GN Broth
Form Powder
Media Type GN Broth
Product Type Nutrient Broth
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.