Learn More
Thermo Scientific™ GN Broth (Dehydrated)
Catalog No. p-6097775
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453514 2.5 kg
R453512 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R453514 Supplier Thermo Scientific™ Supplier No. R453514
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selectively enrich gram-negative bacilli, especially Salmonella and Shigella spp. using GN Broth.
Selectively enrich gram-negative bacilli, especially Salmonella and Shigella spp. using Thermo Scientific™ Remel™ GN Broth (Dehydrated). It was developed by Hanja as an enrichment medium which has proven superior to direct inoculation of media for recovery and enrichment of enteric gram-negative bacilli1. Taylor and Schelhart found GN Broth to be more effective than selenite broth for recovery of Shigella spp2.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.
Selectively enrichgram-negative bacilli, especially Salmonella and Shigella spp. using GN Broth.
This medium is a ready-to-use liquid medium used for qualitative procedures. This medium contains casein and meat peptone that provide amino acids and other nitrogenous compounds to support bacterial growth. Dextrose and mannitol are energy sources and their concentrations are balanced to limit the growth of Proteus and encourage the growth of Salmonella and Shigella. Sodium deoxycholate and sodium citrate inhibit gram-positive organisms.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description GN Broth (Hajna)
Form Powder
Product Type Dehydrated Culture Media
Quantity 2.5 kg