Learn More
Thermo Scientific™ Remel™ Gram Enhancer
Catalog No. R40232
Change view
Click to view available options
Quantity:
250mL/Bottle
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R40232 Each
R40231 250mL/Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R40232 Supplier Thermo Scientific™ Supplier No. R40232
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms.
Thermo Scientific™ Remel Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
Volume (Metric) 3.785 L
Product Type Enhancer