Learn More
Thermo Scientific™ Remel™ Gram Safranin
GSA_VA
Catalog No. R40058
Change view
Click to view available options
Volume (Metric):
250 mL
3.785 L
5 x 250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
R40058 250 mL
R40059 5 x 250 mL
R40079 3.785 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R40058 Supplier Thermo Scientific™ Supplier No. R40058
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
Certifications/Compliance CE Marked
Volume (Metric) 250 mL
Product Type Safranin