Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Haemophilus Test Medium Broth (HTM Broth)

Catalog No. R07681
Change view
Click to view available options
Quantity:
500 mL
20 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R07681 20 x 10 mL
R112380 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R07681 Supplier Thermo Scientific™ Supplier No. R07681
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use for antimicrobial minimal inhibitory concentration (MIC) determinations of Haemophilus spp.

Thermo Scientific™ Remel Haemophilus Test Medium Broth (HTM Broth) is for the antimicrobial minimal inhibitory concentration (MIC) determinations of Haemophilus spp.

TRUSTED_SUSTAINABILITY

Specifications

Description Haemophilus Test Medium Broth
Format 16mm x 125mm Tube
Product Type Microbiology Media
Quantity 20 x 10 mL

REF R112380 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.