Learn More
Thermo Scientific™ Remel™ HC Agar
Catalog No. R453532 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453532 500 g
1 options
This item is not returnable. View return policy
Catalog No. R453532 Supplier Thermo Scientific™ Supplier No. R453532
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cultivate and enumerate molds in cosmetic products using HC Agar Base (Dehydrated) supplemented with polysorbate 80.
Cultivate and enumerate molds in cosmetic products using Thermo Scientific™ Remel™ HC Agar Base (Dehydrated) supplemented with polysorbate 80. HC Agar is prepared by addition of polysorbate 80 to HC Agar base. In 1986, Mead and O'Neill described HC Agar for enumerating molds in cosmetic products1. Polysorbate 80 is used to neutralize residual amounts of preservatives and sequesters surfactants that may be present in the sample.
Cosmetics do not need to be sterile, but they must be adequately preserved. Microbial contamination to cosmetics is a substantial risk to product quality, regulatory compliance and consumer health2. HC Agar Base, formulated by Mead and ONeill, is used for enumerating molds in cosmetic products1. This medium differs from the traditionally used media for testing cosmetics products by addition of Polysorbate 80 and incubation time of 3 days, rather than 7 days, at 27°C ± 0.5°C to obtain a significant mold count3.
  • Selective medium - Chloramphenicol inhibits bacteria, including Pseudomonas aeruginosa and Serratia marcescens, that are potential contaminants of cosmetic products.
The medium contains casein and meat peptones that supply essential amino acids, peptides, and nutrients necessary for microbial growth. Dextrose is the source of carbohydrate energy. Yeast extract supplies B-complex vitamins as well as serves as a growth enhancer. Essential ions are provided by ammonium chloride and magnesium sulfate. Sodium carbonate inactivates low levels of preservatives. Chloramphenicol inhibits bacteria, including Pseudomonas aeruginosa and Serratia marcescens.
TRUSTED_SUSTAINABILITY
Specifications
Description Hungate Cap Agar
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only