Description
Remel Lactose Broth (Dehydrated) is used for detection of coliform bacteria in water, foods, and dairy products according to standard methods.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, EPA, USDA, USP. |
| Description | Lactose Broth |
| Form | Powder |
| Media Type | Lactose Broth |
| Product Type | Nutrient Broth |
| Quantity | 2.5 kg |
Not intended for IVD use.