Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Lactose Broth (Dehydrated)

Catalog No. R453656 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453656 10 kg
R453654 2.5 kg
R453652 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R453656 Supplier Thermo Scientific™ Supplier No. R453656
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Remel Lactose Broth (Dehydrated) is used for detection of coliform bacteria in water, foods, and dairy products according to standard methods.

Remel Lactose Broth (Dehydrated) is used for detection of coliform bacteria in water, foods, and dairy products according to standard methods.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, EPA, USDA, USP.
Description Lactose Broth
Form Powder
Media Type Lactose Broth
Product Type Nutrient Broth
Quantity 10 kg

Not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.