Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lauryl Tryptose Broth (Dehydrated)

Catalog No. R453664 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453664 2.5 kg
R453662 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R453664 Supplier Thermo Scientific™ Supplier No. R453664
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect coliform organisms in water, wastewater and foods with Lauryl Tryptose Broth.

Detect coliform organisms in water, wastewater and foods with Thermo Scientific™ Remel™ Lauryl Tryptose Broth (Dehydrated). Mallman and Darby1-2 evaluated a variety of wetting agents and determined sodium lauryl sulfate added as a selective agent produced the best results for the detection of coliform group. It comes with an inverted Durhams tube which traps the gas which in turn serves as a presumptive test for the presence of coliforms. This medium is recommended by the American Public Health Association, AOAC International (AOAC) and the U.S. Food and Drug Administration3-6.

Detect coliform organisms in water, wastewater and foods with Lauryl Tryptose Broth. A positive fermentation reaction is indicated by turbidity in the broth and gas bubbles in the inverted Durhams tube.

  • Ease of Identification: Gas formation observed in the inverted Durhams Tube by coliform organisms.
  • Selective: Sodium lauryl sulfate is a selective agent that inhibits many organisms other than coliforms.

This medium is used for qualitative procedures. This medium contains casein peptones that provides nitrogen, carbon and sulfur essential for bacterial growth. Lactose is the carbohydrate source fermented by coliforms. Sodium lauryl sulfate is a selective agent that inhibits many organisms other than coliforms.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Lauryl Tryptose Broth
Form Powder
Media Type Lauryl Tryptose Broth
Product Type Nutrient Broth
Quantity 2.5 kg

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.