Learn More
Thermo Scientific™ Remel™ LB Broth, Lennox
Catalog No. R452332 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R452332 500 g
1 options
This item is not returnable. View return policy
Catalog No. R452332 Supplier Thermo Scientific™ Supplier No. R452332
Add to Cart
Add to Cart
This item is not returnable. View return policy
Standard bacteriological nutrient media for cultivation and maintenance of Escherichia coli.
Remel LB Broth, Lennox (Luria broth, Lennox formulation) is used as a standard bacteriological nutrient media for cultivation and maintenance of Escherichia coli.
TRUSTED_SUSTAINABILITY
Specifications
Description LB Broth, Lennox
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
For Laboratory Use Only