Learn More
Thermo Scientific™ Letheen Agar Modified (Dehydrated)
Catalog No. R453722 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453722 500 g
1 options
Catalog No. R453722 Supplier Thermo Scientific™ Supplier No. R453722
Add to Cart
Add to Cart
Effectively carry out microbiological testing of cosmetics using Letheen Agar Modified.
Effectively carry out microbiological testing of cosmetics using Thermo Scientific™ Remel™ Letheen Agar Modified (Dehydrated). Weber and Black recommended the use of lecithin and polysorbate 80 to neutralize the antimicrobial action of quaternary ammonium compounds1,2. Letheen Agar Modified is based on the formula in the Bacteriological Analytical Manual for use in the microbiological testing of cosmetics3. It is recommended by the Association of Official Analytical Chemists (AOAC) for cultivation of microorganisms from cosmetic sampling4.
Effectively carry out microbiological testing of cosmetics using Letheen Agar Modified.
  • Recommended by AOAC for cultivation of microorganisms from cosmetic samples.
  • Based on the formula in in the Bacteriological Analytical Manual for use in the microbiological testing of cosmetics3.
This medium contains casein peptone and beef extract which supply nutrients necessary for the growth of bacteria. Letheen agar modified contains additional casein peptone which supplies nitrogen for enhanced growth of microorganisms. Yeast extract provides B-complex vitamins. Sodium bisulfite neutralizes quaternary ammonium compounds. Lecithin and polysorbate 80 partially inactivate quaternary ammonium compounds, phenolic compounds, formalin, hexachlorophene and ethanol. Dextrose is a carbon source.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Letheen Agar Modified
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only