Description
Remel Letheen Broth (Dehydrated) is used for testing quaternary ammonium compounds for antimicrobial activity.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AOAC. |
| Description | Letheen Broth |
| Form | Powder |
| Media Type | Letheen Broth |
| Product Type | Nutrient Broth |
| Quantity | 500 g |
REF R07700, REF R112420, REF R112423, and REF R117259 not intended for IVD use.