Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Letheen Broth (Dehydrated)

Catalog No. R453714 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453714 2.5 kg
R453712 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R453714 Supplier Thermo Scientific™ Supplier No. R453714
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Remel Letheen Broth (Dehydrated) is used for testing quaternary ammonium compounds for antimicrobial activity.

Remel Letheen Broth (Dehydrated) is used for testing quaternary ammonium compounds for antimicrobial activity.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC.
Description Letheen Broth
Form Powder
Media Type Letheen Broth
Product Type Nutrient Broth
Quantity 2.5 kg

REF R07700, REF R112420, REF R112423, and REF R117259 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.