Learn More
Thermo Scientific™ Letheen Broth Modified (Dehydrated)
Catalog No. R453732 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453732 500 g
1 options
This item is not returnable. View return policy
Catalog No. R453732 Supplier Thermo Scientific™ Supplier No. R453732
Add to Cart
Add to Cart
This item is not returnable. View return policy
Effectively carry out microbiological testing of cosmetics using Letheen Broth Modified.
Effectively carry out microbiological testing of cosmetics using Thermo Scientific™ Remel™ Letheen Broth Modified (Dehydrated). Weber and Black modified Tryptose Glucose Extract Broth by incorporating lecithin and polysorbate 80 as a result of their study on quaternary ammonium compounds1,2. Letheen Broth Modified is based on the formula in the Bacteriological Analytical Manual for use in the microbiological testing of cosmetics3. It is recommended by the Association of Official Analytical Chemists (AOAC) for cultivation of microorganisms from cosmetic samples4.
Effectively carry out microbiological testing of cosmetics using Letheen Broth Modified.
  • Recommended by AOAC for cultivation of microorganisms from cosmetic samples4. 
  • Based on the formula in the Bacteriological Analytical Manual for use in the microbiological testing of cosmetics3.
This medium contains meat peptone and beef extract which supply nutrition necessary for the growth of bacteria. Letheen broth modified contains additional casein peptone which supplies nitrogen for enhanced growth of microorganisms. Yeast extract provides B-complex vitamins. Sodium bisulfite neutralizes quaternary ammonium compounds, lecithin and polysorbate 80 partially inactivate quaternary ammonium compounds, phenolic compounds, formalin, hexachlorophene and ethanol.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Letheen Broth Modified
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
For Laboratory Use Only