Learn More
Thermo Scientific™ Remel™ Levine EMB Agar, Dehydrated
Catalog No. R453402 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453402 500 g
1 options
Catalog No. R453402 Supplier Thermo Scientific™ Supplier No. R453402
Add to Cart
Add to Cart
Remel EMB Agar, Levine (Dehydrated) provides selective isolation and differentiation of gram-negative enteric bacilli from clinical and industrial samples.
Remel EMB Agar, Levine (Dehydrated) provides selective isolation and differentiation of gram-negative enteric bacilli from clinical and industrial samples.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM, COMPF, IDF, USP.
Description EMB Agar, Levine
Form Powder
Product Type Agar
Quantity 500 g