Learn More
Thermo Scientific™ L-J Gruft
Catalog No. R10103
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
1 oz. Bottle
20mm x 113mm Tube - Slant
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R10103 10/Pk. 1 oz. Bottle
R08520 100/Pk. 20mm x 113mm Tube - Slant
R08522 20/Pk. 20mm x 113mm Tube - Slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R10103 Supplier Thermo Scientific™ Supplier No. R10103
This item is not returnable. View return policy
Selectively isolate and cultivate Mycobacterium spp. using Lowenstein-Jensen (L-J) Medium, L-J Gruft, and L-J Mycobactocel.
Selectively isolate and cultivate Mycobacterium spp. using Thermo Scientific™ Remel™ Lowenstein-Jensen (L-J) Medium, L-J Gruft, and L-J Mycobactocel. Medium consisting of congo red and malachite green dyes was formulated by Lowenstein to cultivate mycobacteria1. This medium was modified by Jensen by replacing congo red with citrate and phosphate contents and increasing the malachite green concentration2. L-J Medium used for recovery of pathogenic Mycobacterium spp. was found to be superior to other media tested, including ATS (American Trudeau Society) Medium3,4.
Selectively isolate and cultivate Mycobacterium spp. using Lowenstein-Jensen (L-J) Medium, L-J Gruft, and L-J Mycobactocel.
  • Selective – Antibiotics in the medium make the medium selective for Mycobacterium
TRUSTED_SUSTAINABILITY
Specifications
Description Lowenstein-Jensen Gruft. (LJ) w/penicillin, nalidixic acid
Format 1 oz. Bottle
Product Type Prepared Microbiology Tubes
Quantity 10/Pk.