Learn More
Thermo Scientific™ Remel™ Malonate Broth Modified
Catalog No. R453882 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453882 500 g
1 options
Catalog No. R453882 Supplier Thermo Scientific™ Supplier No. R453882
The Remel Malonate Broth Modified is used for the differentiation of Enterobacteriaceae on the basis of malonate utilization.
Remel Malonate Broth Modified is used for the differentiation of Enterobacteriaceae on the basis of malonate utilization.
TRUSTED_SUSTAINABILITY
Specifications
Description Malonate Broth Modified
Format Powder
Quantity 500 g
For Laboratory Use Only