Learn More
Thermo Scientific™ Malt Extract Agar (Dehydrated)
Catalog No. R453892 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453892 500 g
1 options
This item is not returnable. View return policy
Catalog No. R453892 Supplier Thermo Scientific™ Supplier No. R453892
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate and enumerate yeast and molds from dairy products and other foodstuffs using Malt Extract Agar
Isolate and enumerate yeast and molds using Thermo Scientific™ Remel™ Malt Extract Agar (Dehydrated). The utilization of malt or malt extracts for the propagation of yeast and molds is quite common. Reddish described the use of malt extract as a substitute for beer wort in a culture medium1. Thom and Church developed Malt Extract Agar, a modification of Reddish's formula, as a general cultivation medium for aspergilli2. Malt Extract Agar is recommended for the detection of yeasts and molds in dairy products and other foodstuffs3-5.
Isolate and enumerate yeast and molds in dairy products and other foodstuffs using Malt Extract Agar. Both fresh and processed products are subject to fungal spoilage and even heat-processed foods may be affected. Certain foodborne molds and yeasts may also elicit allergic reactions or may cause infections.
This medium contains high concentration of maltose which makes it especially suitable for the growth of yeasts and molds. Dextrin and glycerol are carbon sources and gelatin peptone is a source of nitrogen. The acidic pH of this medium is optimum for the growth of yeasts and molds while inhibiting the growth of bacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial reference: COMPF.
Description Malt Extract Agar
Form Powder
Product Type Agar
Quantity 500 g
Not intended for IVD use.