Learn More
Thermo Scientific™ Remel™ Microloop
Description
Thermo Scientific™ Remel Microloop is a twisted nichrome loop without handle.
Specifications
Specifications
| Volume | 0.01 μL |
| Material | Nichrome |
| Size | 1/100 |
| Quantity | 1 Ea. |
| Calibrated | Yes |