Learn More
Thermo Scientific™ Middlebrook 7H10 Agar (Dehydrated)
Catalog No. R453982 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453982 500 g
1 options
This item is not returnable. View return policy
Catalog No. R453982 Supplier Thermo Scientific™ Supplier No. R453982
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate and cultivate Mycobacterium spp. using Middlebrook 7H10 Agar.
Isolate and cultivate Mycobacterium spp. using Thermo Scientific™ Remel™ Middlebrook 7H10 Agar (Dehydrated). Dubos and Middlebrook developed media formulations containing oleic acid and albumin which enhanced the growth of tubercle bacilli and protected the organisms against a variety of toxic agents1. In 1958, Middlebrook and Cohn improved the first formulation of oleic acid and albumin agar to obtain 7H10 Agar, which allows faster and more luxuriant growth of Mycobacterium species2.
Isolate and cultivate Mycobacterium spp. using Middlebrook 7H10 Agar.
  • Selective: For Mycobacterium spp from other bacteria due to the presence of malachite-green dye.
  • Allows luxuriant growth of Mycobacterium
This medium is used for qualitative procedures. This medium contains inorganic salts which are essential for the growth of mycobacteria. Malachite-green dye acts as a selective agent which partially inhibits bacteria other than mycobacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Middlebrook 7H10 Agar
Form Powder
Product Type Agar
Quantity 500 g
REF R09565 requires addition of OADC enrichment, REF R450600.