Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Middlebrook 7H9 Broth (Dehydrated)

Catalog No. R454014 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R454014 2.5 kg
R454012 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R454014 Supplier Thermo Scientific™ Supplier No. R454014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and cultivate Mycobacterium spp. using Middlebrook 7H9 Broth.

Isolate and cultivate Mycobacterium spp. using Thermo Scientific™ Remel™ Middlebrook 7H9 Broth (Dehydrated). In 1958, Middlebrook and Cohn were the first to formulate this medium for the cultivation of mycobacteria1. This medium is found useful in the transfer of cultures to be tested with biochemical media2.

Isolate and cultivate Mycobacterium spp. using Middlebrook 7H9 Broth.

  • Ease of Handling: Ease of transfer of cultures to be tested with biochemical media.
  • Allows luxuriant growth of Mycobacterium

This medium is used for qualitative procedures. This medium contains inorganic salts which are essential for the growth of mycobacteria. Ammonium sulphate acts as a source of nitrogen. Biotin plays an important role in carboxylation and decarboxylation reactions, while pyridoxin is a precursor of enzymatic activity. The addition of ADC Enrichment provides a carbon source of energy, and catalase minimizes toxic effects.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Middlebrook 7H9 Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 2.5 kg

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.