Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Middlebrook 7H9 Broth w/Polysorbate 80

Catalog No. R09556
Change view
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R09556 100 x 5 mL
R09558 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R09556 Supplier Thermo Scientific™ Supplier No. R09556
Add to Cart
Add to Cart
This item is not returnable. View return policy

Differentiate and identify Mycobacterium species based on their ability to reduce potassium tellurite using Middlebrook 7H9 Broth w/Polysorbate 80.

Differentiate Mycobacterium species based on their ability to reduce potassium tellurite using Thermo Scientific™ Remel™ Middlebrook 7H9 Broth w/Polysorbate 80 (Tellurite Reduction Medium). Middlebrook and Cohn developed Middlebrook 7H9 Broth in 1958 to cultivate mycobacteria1. Kilburn et al. described the differential identification of mycobacteria using the tellurite reduction test2. They reported that all mycobacteria growing actively on a solid medium rapidly reduced a solution of tellurite salt used to flood the plate.

Requires:

Requires Potassium Tellurite Reagent, REF R21254.

TRUSTED_SUSTAINABILITY

Specifications

Description Middlebrook 7H9 Broth with Polysorbate 80 Tellurite Reduction Medium
Format 20mm x 150mm Tube
Product Type Microbiology Media
Quantity 100 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.