Learn More
Thermo Scientific™ MRS Agar (Dehydrated)
Catalog No. R454052 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R454052 500 g
1 options
This item is not returnable. View return policy
Catalog No. R454052 Supplier Thermo Scientific™ Supplier No. R454052
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Isolate and enumerate Lactobacillus species and gram-positive cocci using MRS Agar.
Isolate and enumerate Lactobacillus species and gram-positive cocci from clinical and non-clinical samples using Thermo Scientific™ Remel™ MRS Agar (Dehydrated). In 1960, De Man, Rogosa and Sharpe modified a medium earlier formulated by Rogosa et al1 by eliminating tomato juice and created a medium which supported the growth of slower growing lactobacilli2. Recently MRS Agar has been used in the clinical laboratory to differentiate certain strains of gram-positive cocci and lactobacilli from clinical samples3-6.
‘Lactic acid bacteria’ are gram-positive, catalase and oxidase negative and are fastidious in their nutritional requirements. Growth is enhanced considerably by microaerobic conditions. Generally, the ‘lactic acid bacteria’ show delayed growth and smaller colony size than other microorganisms. They may be overgrown in non-selective media, especially if incubation is required for 2-4 days.
MRS Agar is designed to encourage the growth of the ‘lactic acid bacteria’ which includes species of the following genera: Lactobacillus, Streptococcus, Pediococcus, and Leuconostoc. All these species can produce lactic acid in considerable amounts.
  • Can be used to differentiate certain strains of gram-positive cocci and lactobacilli.
  • Supports the growth of slower growing lactobacilli.
  • Low Selectivity: ammonium citrate and sodium acetate inhibit the growth of commensal microbial flora, gram-negative bacilli, and fungi, and improve the growth of lactobacilli.
This medium contains gelatin peptone and beef extract that supply essential nutrients and amino acids necessary for bacterial growth. Yeast extract is the growth enhancer and dextrose provide an energy source. Polysorbate 80 supplies fatty acids required for the metabolism of lactobacilli. Ammonium citrate and sodium acetate inhibit the growth of commensal microbial flora, gram-negative bacilli, and fungi, and improve the growth of lactobacilli.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description MRS Agar
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only