Learn More
Thermo Scientific™ MRS Broth (Dehydrated)
Catalog No. R454062 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R454062 500 g
1 options
This item is not returnable. View return policy
Catalog No. R454062 Supplier Thermo Scientific™ Supplier No. R454062
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate Lactobacillus species and differentiate gram-positive cocci using MRS Broth.
Isolate Lactobacillus species and differentiate gram-positive cocci using Thermo Scientific™ Remel™ MRS Broth (Dehydrated). In 1960, De Man, Rogosa and Sharpe modified a medium formulated earlier by Rogosa et al1 by eliminating tomato juice and created a medium which supported the growth of slower growing lactobacilli2. MRS broth is named for de Man, Rogosa and Sharpe and has been used to differentiate certain strains of gram-positive cocci based on their ability to produce gas in this medium.3-6.
Lactic acid bacteria are Gram-positive, catalase- and oxidase-negative and are fastidious in their nutritional requirements. Growth is enhanced considerably by microaerobic conditions. Generally, the lactic acid bacteria show delayed growth and smaller colony size than other microorganisms. They may be overgrown in non-selective media, especially if incubation is required for 2-4 days.
MRS Agar is designed to encourage the growth of lactic acid bacteria, which includes species of the following genera: Lactobacillus, Streptococcus, Pediococcus, and Leuconostoc. All these species can produce lactic acid in considerable amounts.
  • Can be used to differentiate certain strains of gram-positive cocci.
  • Supports the growth of slower growing lactobacilli.
  • Ammonium citrate and sodium acetate inhibit the growth of certain microorganisms, gram-negative bacteria, and molds.
This medium contains gelatin peptone and beef extract that supply essential nutrients and amino acids necessary for growth of lactobacilli. Yeast extract is a source of B-complex vitamins and dextrose provides an energy source. Polysorbate 80 supplies fatty acids required for the metabolism of lactobacilli. Ammonium citrate and sodium acetate inhibit the growth of certain microorganisms, gram-negative bacteria, and molds.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description MRS Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
For Laboratory Use Only