Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mueller Hinton Agar with 5% Sheep Blood

Catalog No. R04055
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
40/Pk.
Format:
15mm x 100mm Plate
15mm x 150mm Plate
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R04055 10/Pk. 15mm x 150mm Plate
R01622 10/Pk. 15mm x 100mm Plate
R01623 100/Pk. 15mm x 100mm Plate
R04057 40/Pk. 15mm x 150mm Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R04055 Supplier Thermo Scientific™ Supplier No. R04055
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Perform effortless Antimicrobial Susceptibility Tests (AST) for fastidious organisms using this medium recommended by CLSI.

Perform effortless Antimicrobial Susceptibility Tests (AST) for fastidious organisms using Thermo Scientific™ Remel™ Mueller Hinton Agar with 5% Sheep Blood prepared media plates. A precise clear zone will help to determine if the bacteria is resistant or sensitive to the antibiotics used. Mueller-Hinton Agar with 5% Sheep Blood conforms to the American standard of the Clinical and Laboratory Standards Institute (CLSI)1.

The major use of Mueller-Hinton Agar with 2% Salt is for Antimicrobial Susceptibility Testing. The medium is designed and manufactured according to CLSI standards and can be used in internationally recognized standard procedures. 

  • Recommended by the Clinical and Laboratory Standards Institute (CLSI) for routine susceptibility testing of non-fastidious bacteria1.
  • Ready-to-use convenience of prepared media.

Mueller Hinton Agar contains beef extract and acid hydrolysate of casein which supply amino acids, nitrogenous substances, vitamins, and minerals necessary for growth. The starch present acts as a protective colloid against toxic materials present in the medium. The medium contains low levels of thymidine and thymine, as excess amounts can reverse the inhibitory effect of sulphonamides and trimethoprim. The calcium and magnesium levels are controlled so appropriate activity of aminoglycosides, tetracycline, and colistin can be expected when testing Pseudomonas aeruginosa. Agar is the solidifying agent. Sheep blood is added to enhance the growth of Streptococcus pneumoniae.

TRUSTED_SUSTAINABILITY

Specifications

Description Mueller Hinton Agar w/5% Sheep Blood
Format 15mm x 150mm Plate
Product Type Agar Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.