Learn More
Thermo Scientific™ Remel™ NAC Attack
Catalog No. R210250
Change view
Click to view available options
Quantity:
5 x 50 mL
5 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R210250 5 x 50 mL
R210500 5 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R210250 Supplier Thermo Scientific™ Supplier No. R210250
Add to Cart
Add to Cart
This item is not returnable. View return policy
Remel NAC Attack is for digestion and decontamination procedures for AFB cultures.
Remel NAC Attack is for digestion and decontamination procedures for AFB cultures.
Includes:
Five sealed ampules of lyophilized N-acetyl-L-cysteine; five bottles TB Base Digestant.
TRUSTED_SUSTAINABILITY
Specifications
Description NAC Attack
Product Type Bacterial and Fungal Isolation
Quantity 5 x 50 mL