Learn More
Thermo Scientific™ Remel™ PathoDxtra™ Strep Grouping Kit
Description
Eliminate the need for incubation and provide a faster strep grouping solution than more traditional enzyme extraction with the Thermo Scientific™ PathoDxtra™ Strep Grouping Kit.
- Improves workflow efficiencies — Instant acid extraction without the need for incubation or reconstitution
- Saves time — Simple, rapid procedure produces easy-to-read results
- Reduces costs — Identifies all clinically important Strep groups, including Group D; no additional reagents required
- Provides flexibility — Pack variants available to suit testing needs
Includes:
PathoDxtra Strep Grouping Kit (Mfr. No. DR0700M): 3mL each of A, B, C, D, F and G grouping latex; Polyvalent Positive Control, 2.8mL; Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; Mixing Sticks; Disposable Reaction Cards
PathoDxtra Extraction Reagent Set (Mfr. No. DR0709M): Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; For use with the PathoDxtra Strep Grouping Kit
PathoDxtra Strep Grouping Reagent Set (Mfr. No. DR0710M): Extraction Reagent 1 - 4mL; Extraction Reagent 2 - 4mL; Extraction Reagent 3 - 2 x 10mL; Mixing Sticks, Disposable Reaction Cards; For use with selected, separately purchased PathoDxtra Strep Grouping latex reagents
Specifications
Specifications
| Control Sets | Positive |
| Description | Strep Group B Latex |
| Detectable Analytes | beta-haemolytic Streptococci |
| Type | Streptococcus Test Kit |
| For Use With (Application) | Enzyme Immunoassay Detection |
| Material | Polyvalent |
| Organism Group | Group A, B, C, D, F and G |
| Quantity | 60 Tests |
Rapid Streptococcal grouping tests correlate best with reference methods when only Streptococci that are Β-hemolytic on sheep blood agar are tested. 1, 2, 3, 4
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.