Learn More
Thermo Scientific™ Phenol Red Broth Base (Dehydrated)
Catalog No. R454272 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R454272 500 g
1 options
Catalog No. R454272 Supplier Thermo Scientific™ Supplier No. R454272
Add to Cart
Add to Cart
Determine fermentation reactions of bacteria using Phenol Red Broth Base.
Determine fermentation reactions of bacteria with Thermo Scientific™ Remel™ Phenol Red Broth Base (Dehydrated). Vera recommended using casein peptone in fermentation test media because of its high growth-promoting qualities1. It was reported that peptone could be used with phenol red as the pH indicator in fermentation tests.
Phenol Red Broth Base is a basal medium prepared with phenol red as an indicator which changes the medium to yellow when acid is produced. Carbohydrates may be added to the broth base at a final concentration of 1%. A Phenol Red Broth Base control tube is used as a negative control for fermentation studies. A Durham tube may be used to allow detection of gas production.
  • Can be used in fermentation studies of pure cultures of many species of bacteria due to high growth-promoting qualities of casein peptone.
  • Recommended by Association of Official Analytical Collaboration (AOAC) International and the American Public Health Association (APHA)2-5
  • Easy to interpret because of fermentation reaction and gas production. The medium is yellow when there is acid production and red-orange color when there is no fermentation. Bubbles present in the Durham tube indicate gas production.
Phenol Red Broth Base contains casein peptone that has high growth-promoting qualities. Phenol red is the indicator which determines the reaction by a color change. Sodium chloride supplies essential electrolytes and maintains osmotic equilibrium of the medium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Phenol Red Broth Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
For Laboratory Use Only