Learn More
Thermo Scientific™ Remel™ Phenylethyl Alcohol Agar
Catalog No. R454302 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R454302 500 g
1 options
This item is not returnable. View return policy
Catalog No. R454302 Supplier Thermo Scientific™ Supplier No. R454302
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selectively isolate gram-positive cocci using Phenylethyl Alcohol Agar (PEA) (Dehydrated).
Selectively isolate gram-positive cocci using Thermo Scientific™ Remel™ Phenylethyl Alcohol Agar (PEA) (Dehydrated). Phenylethyl Alcohol Agar was formulated by Lilley and Brewer to isolate gram-positive cocci such as staphylococci and streptococci from specimens containing mixed flora1. Phenylethyl alcohol is bacteriostatic and acts on gram-negative bacteria by inhibiting DNA synthesis. The addition of sheep blood provides many growth factors.
Phenylethyl Alcohol Agar allows the growth of gram-positive organisms and inhibits most gram-negative organisms, especially the swarming of Proteus species.
The medium contains casein peptone which provides nitrogen, carbon, and sulfur nutrients necessary for bacterial growth. Sodium chloride supplies essential electrolytes and maintains the osmotic equilibrium. Phenylethyl alcohol inhibits gram-negative organisms by inhibiting DNA synthesis. Sheep blood is added to the medium to provide many growth factors.
TRUSTED_SUSTAINABILITY
Specifications
Description Phenylethyl Alcohol Agar
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only