Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Phosphate Buffer w/MgCl2 (pH 7.2) (Buffered Water) APHA Formulation

Catalog No. R112531
Change view
Click to view available options
Quantity:
100 x 9 mL
10 x 20 mL
Format:
20mm x 113mm Tube
Serum Bottle, Crimp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R112531 10 x 20 mL Serum Bottle, Crimp
R118610 100 x 9 mL 20mm x 113mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R112531 Supplier Thermo Scientific™ Supplier No. R112531
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Thermo Scientific™ Remel Phosphate Buffer with MgCl2 (pH 7.2) APHA Formulation is used as a diluent in microbial limit testing of food and water.

Thermo Scientific™ Remel Phosphate Buffer with MgCl2 (pH 7.2) (Buffered Water) APHA Formulation is used as a diluent in microbial limit testing of food and water.

Ordering Information:

REF R112530 has open crimp seal. REF R112531 has complete crimp seal.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, APHA.
Description Phosphate Buffer w/MgCl
Format Serum Bottle, Crimp
Product Type Buffer Solution
Quantity 10 x 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.