Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Potassium Chloride Solution
GSA_VA

Catalog No. R062621
Change view
Click to view available options
Quantity:
20 x 4.5 mL
100 x 4.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R062621 20 x 4.5 mL
R062620 100 x 4.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R062621 Supplier Thermo Scientific™ Supplier No. R062621
This item is not returnable. View return policy

Potassium Chloride Solution (pH 2.2, 0.2M) is used as a pretreatment to increase yield of Legionella from clinical specimens.

Pretreat the samples to increase yield of Legionella from clinical specimens using Thermo Scientific™ Remel™ Potassium Chloride Solution (pH 2.2, 0.2M). In 1981, Bopp et al. demonstrated the recovery rate of Legionella from environmental water samples was improved by treating samples with acid prior to plating onto selective media1. Buesching et al. demonstrated the recovery of Legionella organisms from clinical specimens was improved by pre-treatment with potassium chloride-hydrochloric acid solution (pH 2.2)2.

Potassium Chloride Solution (pH 2.2, 0.2M) is a liquid medium that is used as a pretreatment to increase yield of Legionella from clinical specimens. Potassium Chloride Solution (pH 2.2, 0.2M) is an acid wash treatment which serves to decontaminate specimens prior to inoculation of media for Legionella cultivation.

Potassium Chloride Solution (pH 2.2, 0.2M) contains hydrochloric acid (0.2 M) and potassium chloride (0.2M) with pH 2.2.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Potassium Chloride Solution
Format 15mm x 103mm Tube
Product Type Buffer Solution
Quantity 20 x 4.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.