Learn More
Thermo Scientific™ Potato Dextrose Agar (Dehydrated)
Catalog No. R454314 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R454314 2.5 kg
R454312 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R454314 Supplier Thermo Scientific™ Supplier No. R454314
Add to Cart
Add to Cart
This item is not returnable. View return policy
Identify, cultivate and enumerate yeast and molds with this medium.
Identify, cultivate and enumerate yeast and molds with Thermo Scientific™ Remel™ Potato Dextrose Agar (Dehydrated).
Recommended by American Public Health Association (APHA) and Association of Official Analytical Chemists (AOAC), for plate count of yeasts and molds in the inspection of dairy products and foods1,2,3. This medium is used to prepare slide culture of fungi to stimulate the spores4,5. It is used to maintain stock cultures of dermatophytes. Rebel and Taplin used this medium to differentiate atypical dermatophytes based on the pigment produced6.
Identify, cultivate and enumerate yeast and molds using the solid medium Potato Dextrose Agar.
  • Recommended by American Public Health Association (APHA) and Association of Official Analytical Chemists (AOAC), for plate count of yeasts and molds in the inspection of dairy products and foods1,2,3.
  • Can be used to maintain stock culture of dermaophytes.
  • Can be used for differentiation of atypical dermatophytes.
The infusion from potatoes provides a nutritious base for luxuriant development of most fungi. Dextrose added to the medium serves as a growth stimulant.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Potato Dextrose Agar
Form Powder
Product Type Agar
Quantity 2.5 kg
For Laboratory Use Only