Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PRAS Cooked Meat Glucose

Catalog No. R05054
Change view
Click to view available options
Quantity:
20 x 7.5 mL
100 x 7.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R05054 100 x 7.5 mL
R05055 20 x 7.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R05054 Supplier Thermo Scientific™ Supplier No. R05054

Cultivate anaerobic bacteria using PRAS Cooked Meat Glucose.

Cultivate anaerobic bacteria using Thermo Scientific™ Remel™ PRAS Cooked Meat Glucose. This medium is an enrichment broth that is recommended by Virginia Polytechnique Institute (VPI) Anaerobe Labortory Manual1. It supports the growth of spore-forming and non-spore-forming obligate anaerobes and has the capacity to initiate growth of bacteria from minute inocula2. Claros et al. found cooked meat medium superior to thioglycollate broth for stock cultures because it maintains the viability of anaerobic bacteria over extended periods of time3. This medium is also referred to as chopped meat medium.

Cultivate anaerobic bacteria using PRAS Cooked Meat Glucose. R.E. Hungate pioneered the development of Pre-reduced Anaerobically Sterilized (PRAS) media, which are processed in a reduced condition and remain reduced up to and after inoculation4.

  • Ready-to-use – Convenience of a prepared medium.
  • Recommended by Virginia Polytechnique institute (VPI) Anaerobe Labortory Manual1.

This medium contains minimal salts, beef heart, peptone, and dextrose which provide amino acids and other nutrients necessary for the growth of anaerobes. Solid meat particles create favorable growth conditions for anaerobes due to the reducing action of sulfhydryl groups from muscle protein5. Cystine provides sulfahydryl groups which bind hydrogen and produce anaerobic conditions. Vitamin K and hemin are essential for the growth of certain anerobes.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description PRAS Cooked Meat Glucose
Format 15mm x 103mm Tube, Hungate Cap
Product Type Microbiology Media
Quantity 100 x 7.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.