Learn More
Thermo Scientific™ Remel™ Rappaport-Vassiliadis R10 Broth
Catalog No. R456092 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R456092 500 g
1 options
Catalog No. R456092 Supplier Thermo Scientific™ Supplier No. R456092
Add to Cart
Add to Cart
Remel Rappaport-Vassiliadis R10 Broth is used for the selective enrichment of Salmonella species from foods.
Remel Rappaport-Vassiliadis R10 Broth is used for the selective enrichment of Salmonella species from foods.
TRUSTED_SUSTAINABILITY
Specifications
Description Rappaport-Vassiliadis R10 Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
For Laboratory Use Only