Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Rappaport-Vassiliadis Salmonella Enrichment Broth MLT

Catalog No. R117662
Change view
Click to view available options
Quantity:
20 x 10 mL
100 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R117662 100 x 10 mL
R117661 20 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R117662 Supplier Thermo Scientific™ Supplier No. R117662
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel Rappaport-Vassiliadis Salmonella Enrichment Broth MLT facilitates selective enrichment of Salmonella spp.

Thermo Scientific™ Remel Rappaport-Vassiliadis Salmonella Enrichment Broth MLT facilitates selective enrichment of Salmonella spp.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: USP.
Description Rappaport-Vassiliadis Salmonella Enrichment Broth MLT
Format 16mm x 125mm Tube
Product Type Microbiology Media
Quantity 100 x 10 mL

Not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.