Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sabouraud Dextrose Agar (Emmons)
GSA_VA

Catalog No. R08722
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube
15mm x 150mm Plate
20mm x 113mm Tube
Monoplate Deep Pour
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R08722 20/Pk. 20mm x 113mm Tube
R01760 10/Pk. Monoplate Deep Pour
R01763 100/Pk. Monoplate Deep Pour
R064420 100/Pk. 15mm x 103mm Tube
R064422 20/Pk. 15mm x 103mm Tube
R08720 100/Pk. 20mm x 113mm Tube
R04070 10/Pk. 15mm x 150mm Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. R08722 Supplier Thermo Scientific™ Supplier No. R08722
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Isolate and cultivate pathogenic yeast and molds from clinical specimens with this selective medium.

Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar (Emmons) prepared media plates. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concertation of dextrose from 40 to 20 g/L2.

Use Sabouraud Dextrose Agar (Emmons) for primary isolation and cultivation of pathogenic yeasts and molds from clinical specimens.

  • Suitable for isolation of pathogenic fungi from a material containing a large amount of other fungi or bacteria.
  • Ready prepared convenience of prepared media.

Casein and meat peptones supply nitrogenous compounds and amino acids necessary for the growth of fungi. Dextrose is an energy source which serves to support the growth of fungi. Chloramphenicol and gentamicin are broad-spectrum antibiotics which inhibit a range of Gram-positive and Gram-negative bacteria. Cycloheximide is inhibitory to rapidly growing saprophytic fungi that could overgrow slow-growing primary pathogens. Penicillin is active against Gram-positive bacteria. Streptomycin inhibits Gram-positive and Gram-negative bacteria.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Sabouraud Dextrose Agar, Emmons
Format 20mm x 113mm Tube
Product Type Agar Plate
Quantity 20/Pk.

REF R112555 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.